missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Combi Safety Storage Cabinets K-UB-90

Safe and approved storage of aggressive and flammable hazardous substances in working areas

Brand:  asecos™ 3035800135363

2179.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 163.04041kg

 View more versions of this product

Product Code. 15924366

  • £2294.37 / Each
Estimated Shipment: 26-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: body made of powder-coated sheet steel
  • User-friendly and safe: door and drawer remain open in any position, interior of the cabinet completely visible — access to all containers, 93° opening of the wing door
  • Safe: drawer fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
  • No unauthorised use: door and drawer lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system, extraction of both compartments through one ventilation connection
  • Certificates
    • Fire resistance 90 minutes
    • Type tested in accordance with EN 14470-1
    • EN 16121/16122 conformity
    • CE compliant
Specifications

Specifications

502 mm
Combi safety storage cabinets
470 mm
Powder-coated plain
1102 mm
157 kg
Light Gray
502 mm
Wing
2
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
1 x 10 liter drawer (sheet steel powder-coated), 2 x pull-out shelf (panel melamine resin-coated/PP)
Safe and approved storage of aggressive and flammable hazardous substances in working areas
381 mm
Steel
502 x 1102 x 601 mm
Left: 381 x 470 x 502 mm, Right: 468 x 440 x 562 mm
Pull-out
601 mm
K90.060.110.050.UB.ST
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers