missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Combi Safety Storage Cabinets K-UB-90
Safe and approved storage of aggressive and flammable hazardous substances in working areas
£2224.94 - £2768.72
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |
---|---|
Depth (Metric) Exterior | 502 mm |
Color | Light Gray |
Material | Steel |
Product Type | Combi safety storage cabinets |
Alert:
For indoor use only
Product Code | Brand | Model | Weight (Metric) | No. of Doors | No. of Shelves | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Weight (Metric) | No. of Doors | No. of Shelves | Price | Quantity & Availability | ||
15924366
|
asecos™
3035800135363 |
K90.060.110.050.UB.ST | 157 kg | 1 | 2 |
£2294.37
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15924376
|
asecos™
3037700134262 |
K90.060.140.050.UB.3T | 194.50 kg | 3 | 2 |
£2768.72
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15954366
|
asecos™
3036500133845 |
K90.060.110.050.UB.2T | 158.50 kg | 2 | 2 |
£2324.05
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15964366
|
asecos™
3036500134261 |
K90.060.110.050.UB.2T | 155.50 kg | 2 | 2 |
£2224.94
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15994366
|
asecos™
3037000135365 |
K90.060.140.050.UB.ST | 191 kg | 1 | 2 |
£2678.62
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Descrizione
Safe and approved storage of aggressive and flammable hazardous substances in working areas
Function / construction
- Robust construction and longevity: body made of powder-coated sheet steel
- User-friendly and safe: door and drawer remain open in any position, interior of the cabinet completely visible — access to all containers, 93° opening of the wing door
- Safe: drawer fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
- No unauthorised use: door and drawer lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system, extraction of both compartments through one ventilation connection
Certificates
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- CE compliant
Specifica
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant | |
Light Gray | |
Combi safety storage cabinets | |
601 mm | |
2 | |
Powder-coated plain |
502 mm | |
Steel | |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
502 mm | |
Pull-out |