missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet Q-PEGASUS-90 Winged Doors

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3000204033635

3422.29 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 431.57520kg

 View more versions of this product

Product Code. 15960716

  • £3602.41 / Each
Estimated Shipment: 10-01-2025
to see stock.

Alert:

For indoor use only


Description

Description

  • Safe and approved storage of flammable hazardous substances in working areas
  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Effortless use: one-hand operation for double-door cabinets (AGT)
  • User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
  • Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications

Specifications

1647 mm
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
522 mm
Steel
615 x 1193 x 1953 mm
522 x 1050 x 1647 mm
Standard
1953 mm
Q90.195.120.WDAC
90 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Warning Yellow doors
Safety storage cabinet
1050 mm
Powder-coated textured
1193 mm
477.50 kg
Anthracite Gray, Warning Yellow
615 mm
Wing, Auto-close
3
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers