missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet Q-PEGASUS-90 Winged Doors
Safe and approved storage of flammable hazardous substances in working areas
£3497.47 - £4770.00
Specifications
Depth (Metric) Exterior | 615 mm |
---|---|
Depth (Metric) Interior | 522 mm |
Height (Metric) Opening | 90 mm |
Material | Steel |
Product Type | Safety storage cabinet |
Alert:
For indoor use only
Product Code | Brand | Configuration | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Description | Price | Quantity & Availability | ||||
15960716
|
asecos™
3000204033635 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow doors |
£3602.41
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15981176
|
asecos™
3004204133622 |
3 x shelf, 1 x perforated insert, 1 x 22 liter bottom collecting sump (sheet steel powder-coated) | Light gray doors |
£3497.47
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15980746
|
asecos™
3000204133635 |
3 x shelf, 1 x perforated insert, 1 x 33 liter bottom collecting sump (sheet steel powder-coated) | Light gray doors |
£3602.41
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15990716
|
asecos™
3000204033640 |
3 x tray shelf, 1 x perforated insert (135 mm high), 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow doors |
£3804.87
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15921186
|
asecos™
3004204133627 |
4 x 19 liter drawer (sheet steel powder-coated) | Light gray doors |
£4139.30
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15981156
|
asecos™
3004204033627 |
4 x 19 liter drawer (sheet steel powder-coated) | Warning Yellow doors |
£4139.30
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13454347
|
asecos™
3004204130053 |
4 x 19 liter drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Light gray doors |
£4526.20
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13474337
|
asecos™
3004204030053 |
4 x 19 liter drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow doors |
£4526.20
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15970756
|
asecos™
3000204133647 |
4 x 29 liter drawer (sheet steel powder-coated) | Light gray doors |
£4244.24
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15940726
|
asecos™
3000204033647 |
4 x 29 liter drawer (sheet steel powder-coated) | Warning Yellow doors |
£4244.24
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15502934
|
asecos™
3000204130015 |
4 x 29 liter drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Light gray doors |
£4770.00
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13454117
|
asecos™
3000204030015 |
4 x 29 liter drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow doors |
£4770.00
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15941186
|
asecos™
3004204133629 |
6 x 19 liter drawer (sheet steel powder-coated) | Light gray doors |
£4582.38
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15960726
|
asecos™
3000204033649 |
6 x 29 liter drawer (sheet steel powder-coated) | Warning Yellow doors |
£4687.32
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Safe and approved storage of flammable hazardous substances in working areas
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Effortless use: one-hand operation for double-door cabinets (AGT)
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Warranty
10 years
Specifications
615 mm | |
90 mm | |
Safety storage cabinet | |
1953 mm | |
2 |
522 mm | |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas | |
1647 mm | |
Powder-coated textured |