missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-PEGASUS-90 with Integrated AGT

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3012300133745

Additional Details : Weight : 331.45920kg

 View more versions of this product

Product Code. 15952896

  • £3129.65 / Each
Estimated Shipment: 07-11-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Effortless use: one-hand operation for double-door cabinets (AGT)
  • User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
  • Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing  locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1072 mm
3 x shelf, 1 x perforated insert (185 mm), 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
524 mm
Steel
616 x 1196 x 1298 mm
524 x 1050 x 1072 mm
Standard
1298 mm
S90.129.120.WDAC
2
One-hand operation for double-door cabinets (AGT)
Safety storage cabinet
1050 mm
Powder-coated plain
1196 mm
345.50 kg
Light Gray
616 mm
Wing, Auto-close
3
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers