missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PEGASUS-90 with Integrated AGT
Safe and approved storage of flammable hazardous substances in working areas
£3081.95 - £3549.94
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Description | One-hand operation for double-door cabinets (AGT) |
Dimensions (D x W x H) | 616 x 1196 x 1298 mm |
Alert:
For indoor use only
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
15932906
|
asecos™
3012300133753 |
2 x drawer (sheet steel powder-coated) | Light Gray |
£3328.40
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15922936
|
asecos™
3012300233753 |
2 x drawer (sheet steel powder-coated) | Yellow |
£3328.40
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15568219
|
asecos™
3012300133744 |
2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£3081.95
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15932926
|
asecos™
3012300233744 |
2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
£3081.95
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15932936
|
asecos™
3012300233754 |
3 x drawer (sheet steel powder-coated) | Warning Yellow |
£3549.94
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15942906
|
asecos™
3012300133754 |
3 x drawer (sheet steel powder-coated) | Light Gray |
£3549.94
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15952896
|
asecos™
3012300133745 |
3 x shelf, 1 x perforated insert (185 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£3129.65
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15942926
|
asecos™
3012300233745 |
3 x shelf, 1 x perforated insert (185 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
£3129.65
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Effortless use: one-hand operation for double-door cabinets (AGT)
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
616 x 1196 x 1298 mm | |
Steel | |
Wing, Auto-close | |
1298 mm | |
S90.129.120.WDAC | |
Powder-coated plain | |
1050 mm |
616 mm | |
One-hand operation for double-door cabinets (AGT) | |
524 x 1050 x 1072 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1072 mm | |
2 | |
1196 mm |