missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX-90 with Winged/Folding Doors
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3020700230218
Code : NEW
Additional Details : Weight : 310.00000kg
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
1742 mm | |
4 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
524 mm | |
Steel | |
Door open arrest system | |
409.50 kg | |
Yellow | |
1968 mm | |
S90.196.090.FWAS | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
Door open arrest system | |
Safety storage cabinet | |
750 mm | |
Powder-coated plain | |
896 mm | |
616 x 896 x 1968 mm | |
524 x 750 x 1742 mm | |
616 mm | |
Wing, Folding | |
2 |