missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX-90 with Winged/Folding Doors
Safe and approved storage of flammable hazardous substances in working areas
£3016.23 - £4298.30
Specifications
Depth (Metric) Exterior | 616 mm |
---|---|
Depth (Metric) Interior | 524 mm |
Dimensions (D x W x H) | 616 x 896 x 1968 mm |
Dimensions (D x W x H) Interior | 524 x 750 x 1742 mm |
Door Style | Wing, Folding |
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
15576855
|
asecos™
3020700133705 |
3 x shelf, 1 x perforated insert (370 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£3016.23
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15586855
|
asecos™
3020700233705 |
3 x shelf, 1 x perforated insert (370 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
£3016.23
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15983056
|
asecos™
3020700133709 |
4 x drawer (sheet steel powder-coated) | Light Gray |
£3658.06
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15923086
|
asecos™
3020700233709 |
4 x drawer (sheet steel powder-coated) | Yellow |
£3658.06
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13444497
|
asecos™
3020700130218 |
4 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
£4298.30
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13404507
|
asecos™
3020700230218 |
4 x drawer, 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
£4298.30
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15903066
|
asecos™
3020700133711 |
6 x drawer (sheet steel powder-coated) | Light Gray |
£4101.14
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15943086
|
asecos™
3020700233711 |
6 x drawer (sheet steel powder-coated) | Warning Yellow |
£4101.14
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
616 mm | |
616 x 896 x 1968 mm | |
Wing, Folding | |
1968 mm | |
Door open arrest system | |
2 | |
896 mm |
524 mm | |
524 x 750 x 1742 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
S90.196.090.FWAS | |
Powder-coated plain | |
750 mm |