missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-PHOENIX-90 with 2 Doors

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3011800133724

Additional Details : Weight : 449.08920kg

 View more versions of this product

Product Code. 15962606

  • £3915.64 / Each
Estimated Shipment: 26-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Space saving folding doors: less required space in front of the cabinet while doors are open
  • User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing  locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1742 mm
Safety storage cabinet
1050 mm
Powder-coated plain
1196 mm
616 x 1196 x 1968 mm
524 x 1050 x 1742 mm
616 mm
Folding
2
4 x drawer
Safe and approved storage of flammable hazardous substances in working areas
524 mm
Steel
Door open arrest system
493 kg
Light Gray
1968 mm
S90.196.120.FDAS
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers