missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX-90 with 2 Doors
Safe and approved storage of flammable hazardous substances in working areas
£3273.81 - £4737.67
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Dimensions (D x W x H) | 616 x 1196 x 1968 mm |
Material | Steel |
Alert:
For indoor use only
Product Code | Brand | Configuration | Color | Width (Metric) Interior | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Color | Width (Metric) Interior | Price | Quantity & Availability | |||
15376778
|
asecos™
3011800130127 |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump | Light Gray | 1050 mm |
£3598.70
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15975946
|
asecos™
3070800133848 |
3 x shelf, 1 x perforated insert (370 mm), 1 x bottom collecting sump, 3 x tray shelf, 1 x bottom collecting sump (PP), Vertical dividing wall | Light Gray | 450 mm |
£4106.97
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15912636
|
asecos™
3011800233715 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump | Yellow | 1050 mm |
£3273.81
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15912606
|
asecos™
3011800133719 |
3 x tray shelf, 1 x perforated insert (365 mm), 1 x bottom collecting sump | Light Gray | 1050 mm |
£3476.27
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15952636
|
asecos™
3011800233719 |
3 x tray shelf, 1 x perforated insert (365 mm), 1 x bottom collecting sump | Warning Yellow | 1050 mm |
£3476.27
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15962606
|
asecos™
3011800133724 |
4 x drawer | Light Gray | 1050 mm |
£3915.64
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15902646
|
asecos™
3011800233724 |
4 x drawer | Yellow | 1050 mm |
£3915.64
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15982606
|
asecos™
3011800133726 |
6 x drawer | Light Gray | 1050 mm |
£4358.72
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15912646
|
asecos™
3011800233726 |
6 x drawer | Yellow | 1050 mm |
£4358.72
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15985946
|
asecos™
3070800133849 |
6 x drawer, 3 x tray shelf, 1 x bottom collecting sump (PP), Vertical dividing wall | Light Gray | 450 mm |
£4737.67
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding doors: less required space in front of the cabinet while doors are open
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
Steel | |
Folding | |
1968 mm | |
Door open arrest system | |
Powder-coated plain |
616 mm | |
616 x 1196 x 1968 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
2 | |
1196 mm |