missing translation for 'onlineSavingsMsg'
Learn More

asecos Type 90 Safety Storage Cabinet V-MOVE-90

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos 3508505135101

Additional Details : Weight : 361.11600kg

 View more versions of this product

Product Code. 15907766

  • £4453.06 / Each
Estimated Shipment: 26-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
  • Electronic opening and closing: the vertical drawers open and close at the push of a button
  • Perfect access: the vertical drawers provide an overview and simple access to the stored containers
  • Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
  • No unauthorised use: vertical drawers lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system

Certificates

  • Fire resistance: 90 minutes
  • Type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

331 mm
77.40 in.
Gypsum
860 x 449 x 1966 mm
33.86 in.
331 x 775 x 1750 mm
814 lb.
5
68.90 in.
Safety Storage Cabinet
17.68 in.
Anthracite Gray
1966 mm
V90.196.045.VDAC:0013
1
775 mm
Textured
(5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated)
369 kg
30.51 in.
860 mm
50 kg
13.03 in.
1750 mm
Safe and approved storage of flammable hazardous substances in working areas
449 mm
Standard
Auto-close
235/250/250/250/285/260 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers