missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Safety Storage Cabinet V-MOVE-90
Safe and approved storage of flammable hazardous substances in working areas
£4288.23 - £6244.46
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (English) Interior | 30.51 in. |
Depth (Metric) Exterior | 860 mm |
Depth (Metric) Interior | 775 mm |
Depth (English) Exterior | 33.86 in. |
Alert:
For indoor use only
Product Code | Brand | Color | No. of Doors | Surface Coating | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | No. of Doors | Surface Coating | Includes | Price | Quantity & Availability | ||
15907766
|
asecos
3508505135101 |
Anthracite Gray | 1 | Textured | (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£4453.06
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15957756
|
asecos
3508504735100 |
Gentian Blue, Anthracite Gray | 1 | Textured | (4) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£4288.23
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15967756
|
asecos
3508504735101 |
Gentian Blue, Anthracite Gray | 1 | Textured | (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£4329.04
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15977776
|
asecos
3508604735102 |
Gentian Blue, Anthracite Gray | 2 | Textured | Right: (4) shelves, (1) perforated insert, (1) bottom collecting sump; Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£5915.86
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15997776
|
asecos
3508604735104 |
Gentian Blue, Anthracite Gray | 2 | Textured | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated); Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump |
£5956.67
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15987776
|
asecos
3508604735103 |
Gentian Blue, Anthracite Gray | 2 | Textured | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump; Left: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£5997.48
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15947736
|
asecos
3503500135104 |
Light Gray | 2 | Plain | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated); Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump |
£5956.67
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15917756
|
asecos
3508504635100 |
Red, Anthracite Gray | 1 | Textured | (4) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£4288.23
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15927756
|
asecos
3508504635101 |
Red, Anthracite Gray | 1 | Textured | (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£4329.04
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15937776
|
asecos
3508604635104 |
Red, Anthracite Gray | 2 | Textured | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated); Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump |
£5956.67
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15927776
|
asecos
3508604635103 |
Red, Anthracite Gray | 2 | Textured | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump; Left: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£5997.48
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15997756
|
asecos
3508505135100 |
Silver, Anthracite Gray | 1 | Textured | (4) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£4412.25
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15937786
|
asecos
3508605135102 |
Silver, Anthracite Gray | 2 | Textured | Right: (4) shelves, (1) perforated insert, (1) bottom collecting sump; Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£6162.84
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15957786
|
asecos
3508605135104 |
Silver, Anthracite Gray | 2 | Textured | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated); Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump |
£6203.65
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15947786
|
asecos
3508605135103 |
Silver, Anthracite Gray | 2 | Textured | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump; Left: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£6244.46
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15987736
|
asecos
3503500535102 |
White | 2 | Plain | Right: (4) shelves, (1) perforated insert, (1) bottom collecting sump; Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£5915.86
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15907746
|
asecos
3503500535104 |
White | 2 | Plain | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated); Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump |
£5956.67
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15997736
|
asecos
3503500535103 |
White | 2 | Plain | Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump; Left: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£5997.48
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
- Electronic opening and closing: the vertical drawers open and close at the push of a button
- Perfect access: the vertical drawers provide an overview and simple access to the stored containers
- Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
- No unauthorised use: vertical drawers lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
860 mm | |
33.86 in. | |
Gypsum | |
Auto-close | |
68.90 in. | |
1750 mm | |
Standard |
30.51 in. | |
775 mm | |
77.40 in. | |
Safety Storage Cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1966 mm | |
50 kg |