missing translation for 'onlineSavingsMsg'
Learn More

asecos Type 90 Safety Storage Cabinet V-MOVE-90

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos 3508604735104

Additional Details : Weight : 547.92627kg

 View more versions of this product

Product Code. 15997776

  • £5956.67 / Each
Estimated Shipment: 26-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
  • Electronic opening and closing: the vertical drawers open and close at the push of a button
  • Perfect access: the vertical drawers provide an overview and simple access to the stored containers
  • Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
  • No unauthorised use: vertical drawers lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system

Certificates

  • Fire resistance: 90 minutes
  • Type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

701 mm
77.40 in.
Gypsum
860 x 819 x 1966 mm
33.86 in.
701 x 775 x 1750 mm
1219 lb.
9
68.90 in.
Safety Storage Cabinet
32.24 in.
Gentian Blue, Anthracite Gray
1966 mm
V90.196.081.VDAC:0013
2
775 mm
Textured
Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated); Left: (4) shelves, (1) perforated insert, (1) bottom collecting sump
553 kg
30.51 in.
860 mm
50 kg
27.60 in.
1750 mm
Safe and approved storage of flammable hazardous substances in working areas
819 mm
Standard
Auto-close
Right: 235/250/250/250/285/260 mm; Left: 300/285/285/380/290 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers