missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet V-MOVE-90 Anthracite Gray

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3508604135103

Additional Details : Weight : 550.62628kg

 View more versions of this product

Product Code. 15987766

  • £5997.48 / Each
Estimated Shipment: 10-01-2025
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
  • Electronic opening and closing: the vertical drawer opens and closes at the push of a button
  • Perfect access: the vertical drawer provides an overview and simple access to the stored containers
  • Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
  • No unauthorised use: vertical drawer lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1750 mm
5 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), on each side
Safe and approved storage of flammable hazardous substances in working areas
775 mm
Steel
860 x 819 x 1966 mm
775 x 701 x 1750 mm
Standard
1966 mm
V90.196.081.VDAC:0013
2
Light gray vertical drawer doors
Safety storage cabinet
701 mm
Powder-coated textured
819 mm
614 kg
Anthracite Gray, Light Gray
860 mm
Auto-close, Sliding
10
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers