missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet V-MOVE-90 Anthracite Gray
Safe and approved storage of flammable hazardous substances in working areas
£4280.81 - £5998.54
Specifications
Depth (Metric) Exterior | 860 mm |
---|---|
Depth (Metric) Interior | 775 mm |
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Auto-close, Sliding |
Alert:
For indoor use only
Product Code | Brand | No. of Doors | Configuration | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Doors | Configuration | Description | Price | Quantity & Availability | |||
15977746
|
asecos™
3508504135100 |
1 | 4 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light gray vertical drawer doors |
£4288.23
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15957746
|
asecos™
3508504036111 |
1 | 4 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow vertical drawer doors |
£4280.81
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15987746
|
asecos™
3508504135101 |
1 | 5 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Light gray vertical drawer doors |
£4329.04
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15947746
|
asecos™
3508504035101 |
1 | 5 x shelf, 1 x perforated insert, 1 x liter bottom collecting sump (sheet steel powder-coated) | Warning Yellow vertical drawer doors |
£4329.04
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15967746
|
asecos™
3508504036112 |
1 | 5 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow vertical drawer doors |
£4329.57
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15977766
|
asecos™
3508604135102 |
2 | 4 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), on each side | Light gray vertical drawer doors |
£5915.86
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15917766
|
asecos™
3508604035102 |
2 | 4 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), on each side | Warning Yellow vertical drawer doors |
£5915.86
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15947766
|
asecos™
3508604036113 |
2 | 4 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated), on each side | Warning Yellow vertical drawer doors |
£5901.02
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15987766
|
asecos™
3508604135103 |
2 | 5 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), on each side | Light gray vertical drawer doors |
£5997.48
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15927766
|
asecos™
3508604035103 |
2 | 5 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), on each side | Warning Yellow vertical drawer doors |
£5997.48
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15967766
|
asecos™
3508604036115 |
2 | 5 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated), on each side | Warning Yellow vertical drawer doors |
£5998.54
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15937766
|
asecos™
3508604035104 |
2 | 4 x shelf, 1 x perforated insert, 1 x bottom collecting sump, 5 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow vertical drawer doors |
£5956.67
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15957766
|
asecos™
3508604036114 |
2 | 4 x tray shelf, 1 x bottom collecting sump, 5 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow vertical drawer doors |
£5949.78
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
- Electronic opening and closing: the vertical drawer opens and closes at the push of a button
- Perfect access: the vertical drawer provides an overview and simple access to the stored containers
- Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
- No unauthorised use: vertical drawer lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
860 mm | |
Steel | |
Auto-close, Sliding | |
1966 mm | |
Powder-coated textured |
775 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1750 mm |