missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Under Bench Cabinet
Safety storage under bench cabinet
Marque: asecos™ 3048200130488
1426.69 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Informations supplémentaires : Weight : 115.73640kg
Alert:
For indoor use only
Description
- Robust construction and longevity: body made of powder-coated sheet steel
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Spécification
Safety storage under bench cabinet UB-T-90 model UB90.060.059.TR, light gray (RAL 7035), interior equipment with 1 x perforated insert (stainless steel), 1 x bottom collecting sump (stainless steel) | |
Safe and approved storage of flammable hazardous substances in working areas | |
23.35 in. | |
Gypsum | |
Cabinet and interior equipment | |
22.6 in. | |
Light Gray | |
574 mm | |
75 kg | |
UB90.060.059.TR | |
0 | |
1 |
Safety storage under bench cabinet | |
23.62 in. | |
Plain | |
593 mm | |
574 x 593 x 600 mm | |
452 x 470 x 502 mm | |
Without Shelves | |
600 mm | |
Wing | |
593 mm | |
502 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |