missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Under Bench Cabinet
Safety storage under bench cabinet
£1626.04 - £3164.10
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |
---|---|
Color | Light Gray |
Material | Gypsum |
Product Type | Safety storage under bench cabinet |
Door Style | Wing |
Alert:
For indoor use only
Product Code | Brand | Model | No. of Doors | Shelf Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | No. of Doors | Shelf Type | Price | Quantity & Availability | |||
15914556
|
asecos™
3048200130488 |
UB90.060.059.TR | 1 | Without Shelves |
£1626.04
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16307596
|
asecos™
3641800130447 |
UB90.080.110.060.2T | 2 | Without Shelves |
£3121.70
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16317596
|
asecos™
3641800130448 |
UB90.080.110.060.2T | 2 | Shelf |
£3164.10
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16337586
|
asecos™
3591500130485 |
UB90.080.059.060.TR | 1 | Without Shelves |
£1939.80
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16347586
|
asecos™
3591500130486 |
UB90.080.059.060.TR | 1 | Shelf |
£1987.50
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16367576
|
asecos™
3590000130485 |
UB90.080.059.060.T | 1 | Without Shelves |
£1939.80
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16377576
|
asecos™
3590000130486 |
UB90.080.059.060.T | 1 | Shelf |
£1987.50
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: body made of powder-coated sheet steel
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant | |
Gypsum | |
Wing | |
Cabinet and interior equipment |
Light Gray | |
Safety storage under bench cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
Plain |