missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet

Safety storage under bench cabinet

Brand:  asecos™ 3641800130448

Additional Details : Weight : 14.50000kg

 View more versions of this product

Product Code. 16317596

  • £3164.10 / Each
Estimated Shipment: 08-11-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: body made of powder-coated sheet steel
  • Easy handling with comfort: door stops in any position, self-closing in the event of fire
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
  • Fire resistance 90 minutes
  • type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

Safety storage under bench cabinet UB-T-90 model UB90.080.110.060.2T light gray (RAL 7035), interior equipment with 1 x shelf, 1 x perforated insert, 1 x bottom collecting sump (powder-coated sheet steel)
Safe and approved storage of flammable hazardous substances in working areas
43.39 in.
Gypsum
Cabinet and interior equipment
23.62 in.
Light Gray
600 mm
30 kg, 75 kg
UB90.080.110.060.2T
1
2
Safety storage under bench cabinet
31.5 in.
Plain
1102 mm
600 x 1102 x 800 mm
482 x 979 x 702 mm
Shelf
800 mm
Wing
1102 mm
702 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers