missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet

Safety storage under bench cabinet

Brand:  asecos™ 3641800130447

Additional Details : Weight : 14.50000kg

 View more versions of this product

Product Code. 16307596

  • £3121.70 / Each
Estimated Shipment: 26-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: body made of powder-coated sheet steel
  • Easy handling with comfort: door stops in any position, self-closing in the event of fire
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
  • Fire resistance 90 minutes
  • type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

Safety storage under bench cabinet UB-T-90 model UB90.080.110.060.2T light gray (RAL 7035), interior equipment with 1 x perforated insert, 1 x bottom collecting sump (powder-coated sheet steel)
Safe and approved storage of flammable hazardous substances in working areas
43.39 in.
Gypsum
Cabinet and interior equipment
23.62 in.
Light Gray
600 mm
75 kg
UB90.080.110.060.2T
0
2
Safety storage under bench cabinet
31.5 in.
Plain
1102 mm
600 x 1102 x 800 mm
482 x 979 x 702 mm
Without Shelves
800 mm
Wing
1102 mm
702 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers