missing translation for 'onlineSavingsMsg'
Learn More

asecos Type 90 Safety Storage Under Bench Cabinet UB-T-90

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos 3044400133776

2006.60 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 184.00000kg

 View more versions of this product

Product Code. 15928132

  • £2228.32 / Each
Estimated Shipment: 07-11-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: body made of powder-coated sheet steel
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Certificates

  • Fire resistance: 90 minutes
  • Type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

979 mm
23.62 in.
Gypsum
502 x 1102 x 600 mm
19.76 in.
979 x 382 x 502 mm
373 lb.
1
19.76 in.
Under Bench Safety Storage Cabinet
43.39 in.
Light Gray
600 mm
UB90.060.110.050.2T
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
382 mm
Plain
(1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated)
169 kg
15.04 in.
502 mm
30 kg (standard shelf)/75 kg (grid shelf)
38.54 in.
502 mm
Safe and approved storage of flammable hazardous substances in working areas
1102 mm
Standard
Left-hinged, Wing
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers