missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Safety Storage Under Bench Cabinet UB-T-90
Safe and approved storage of flammable hazardous substances in working areas
£1586.18 - £3438.64
Specifications
Color | Light Gray |
---|---|
Material | Gypsum |
Product Type | Under Bench Safety Storage Cabinet |
For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
No. of Shelves | 1 |
Alert:
For indoor use only
Product Code | Brand | Dimensions (D x W x H) | Door Style | No. of Doors | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (D x W x H) | Door Style | No. of Doors | Includes | Price | Quantity & Availability | ||
15938132
|
asecos
3048300133766 |
502 x 593 x 600 mm | Left-hinged, Wing | 1 | (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£1586.18
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15928132
|
asecos
3044400133776 |
502 x 1102 x 600 mm | Left-hinged, Wing | 2 | (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£2228.32
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16387576
|
asecos
3590000130487 |
600 x 593 x 800 mm | Left-hinged, Wing | 1 | (1) drawer max. interior height (sheet steel powder-coated (electrically conductive)) |
£2239.78
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16317586
|
asecos
3590000133763 |
600 x 593 x 800 mm | Left-hinged, Wing | 1 | (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£1764.37
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16397576
|
asecos
3590000130488 |
600 x 593 x 800 mm | Left-hinged, Wing | 1 | (1) perforated insert, (1) bottom collecting sump (stainless steel) |
£1894.22
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16327586
|
asecos
3590000133764 |
600 x 593 x 800 mm | Left-hinged, Wing | 1 | (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£1805.18
Each |
Estimated Shipment: 18-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16307586
|
asecos
3590000130489 |
600 x 593 x 800 mm | Left-hinged, Wing | 1 | (1) shelf, (1) perforated insert (stainless steel), (1) bottom collecting sump (stainless steel) |
£1995.45
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16327596
|
asecos
3641800130449 |
600 x 1102 x 800 mm | Left-hinged, Wing | 2 | (1) drawer max. interior height (sheet steel powder-coated (electrically conductive)) |
£3438.64
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16347596
|
asecos
3641800133771 |
600 x 1102 x 800 mm | Left-hinged, Wing | 2 | (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£2493.12
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16337596
|
asecos
3641800130450 |
600 x 1102 x 800 mm | Left-hinged, Wing | 2 | (1) perforated insert, (1) bottom collecting sump (stainless steel) |
£2690.28
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15951342
|
asecos
3641800130451 |
600 x 1102 x 800 mm | Left-hinged, Wing | 2 | (1) shelf, (1) perforated insert, (1) bottom collecting sump (stainless steel) |
£2819.60
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15985770
|
asecos
3641800133772 |
600 x 1102 x 800 mm | Left-hinged, Wing | 2 | (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£2539.76
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16357586
|
asecos
3591500130487 |
600 x 593 x 800 mm | Right-hinged, Wing | 1 | (1) drawer max. interior height (sheet steel powder-coated (electrically conductive)) |
£2239.78
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16387586
|
asecos
3591500133763 |
600 x 593 x 800 mm | Right-hinged, Wing | 1 | (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£1764.37
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16367586
|
asecos
3591500130488 |
600 x 593 x 800 mm | Right-hinged, Wing | 1 | (1) perforated insert, (1) bottom collecting sump (stainless steel) |
£1894.22
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16397586
|
asecos
3591500133764 |
600 x 593 x 800 mm | Right-hinged, Wing | 1 | (1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) |
£1805.18
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16377586
|
asecos
3591500130489 |
600 x 593 x 800 mm | Right-hinged, Wing | 1 | (1) shelf, (1) perforated insert (stainless steel), (1) bottom collecting sump (stainless steel) |
£1995.45
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Robust construction and longevity: body made of powder-coated sheet steel
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Light Gray | |
Under Bench Safety Storage Cabinet | |
1 |
Gypsum | |
Safe and approved storage of flammable hazardous substances in working areas | |
Plain |