missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Safety Storage Under Bench Cabinet UB-T-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos 3641800133772
2220.30 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00001kg
Alert:
For indoor use only
Description
- Robust construction and longevity: body made of powder-coated sheet steel
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
979 mm | |
31.50 in. | |
Gypsum | |
600 x 1102 x 800 mm | |
23.62 in. | |
979 x 482 x 702 mm | |
476 lb. | |
1 | |
27.64 in. | |
Under Bench Safety Storage Cabinet | |
43.39 in. | |
Light Gray | |
800 mm | |
UB90.080.110.060.2T | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |
482 mm | |
Plain | |
(1) shelf, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) | |
216 kg | |
18.98 in. | |
600 mm | |
30 kg (standard shelf)/75 kg (grid shelf) | |
38.54 in. | |
702 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
1102 mm | |
Standard | |
Left-hinged, Wing | |
2 |