missing translation for 'onlineSavingsMsg'
Learn More

asecos Type 90 Safety Storage Under Bench Cabinet UB-T-90

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos 3641800133771

Additional Details : Weight : 14.50000kg

 View more versions of this product

Product Code. 16347596

  • £2493.12 / Each
Estimated Shipment: 26-11-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: body made of powder-coated sheet steel
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Certificates

  • Fire resistance: 90 minutes
  • Type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

979 mm
31.50 in.
Gypsum
600 x 1102 x 800 mm
23.62 in.
979 x 482 x 702 mm
486 lb.
1
27.64 in.
Under Bench Safety Storage Cabinet
43.39 in.
Light Gray
800 mm
UB90.080.110.060.2T
2
482 mm
Plain
(1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated)
221 kg
18.98 in.
600 mm
75 kg
38.54 in.
702 mm
Safe and approved storage of flammable hazardous substances in working areas
1102 mm
Perforated insert
Left-hinged, Wing
400 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers