missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Type N18 Clear Screw Neck Headspace Vials
Brand: Macherey-Nagel™ 702866
Additional Details : Weight : 0.01000kg
Specifications
Clear | |
10 mL | |
100 | |
Round | |
100 Pk. |
22.5 mm | |
Screw Neck Headspace | |
100 Pack | |
46 mm |