Learn More
DuPont™ Tyvek™ 600 Plus with Socks
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ 600 Plus with Socks is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
£11.75 - £865.00
Specifications
Product Type | Hooded Coveralls |
---|---|
Material | HDPE |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic, Attached Socks, Boot Overflaps |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
11703073
|
DuPont™
D13398935 |
X-Large |
£11.75
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12322259
|
DuPont™
D13398935 |
X-Large |
£227.63
Pack of 20 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
11843510
|
DuPont™
D13398935 |
X-Large |
£840.00
Pack of 80 |
In Stock
Log in to see stock. |
||||||
11833520
|
DuPont™
D13495777 |
Small |
£840.00
Pack of 80 |
In Stock
Log in to see stock. ![]() |
||||||
11773063
|
DuPont™
D13495777 |
Small |
£11.75
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12392249
|
DuPont™
D13495777 |
Small |
£221.00
Pack of 20 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
11843520
|
DuPont™
D13495765 |
Medium |
£840.00
Pack of 80 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12302259
|
DuPont™
D13495765 |
Medium |
£221.00
Pack of 20 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
11793063
|
DuPont™
D13398948 |
Large |
£11.75
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12312259
|
DuPont™
D13398948 |
Large |
£221.00
Pack of 20 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
11833510
|
DuPont™
D13398948 |
Large |
£840.00
Pack of 80 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
11823520
|
DuPont™
D13398927 |
2X-Large |
£840.00
Pack of 80 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
11713073
|
DuPont™
D13398927 |
2X-Large |
£11.75
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12332259
|
DuPont™
D13398927 |
2X-Large |
£221.00
Pack of 20 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
12342259
|
DuPont™
D13495744 |
3X-Large |
£226.00
Pack of 20 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
12623700
|
DuPont™
D13495744 |
3X-Large |
£865.00
Pack of 80 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
Description
- Hooded coverall with integrated socks and additional knee-length boot flap.
- Available in white in sizes S to XXXL.
- Robust yet lightweight (<250g per garment).
- Self-adhesive chin flap for tight seal of suit to the mask.
- Elasticated face, wrists and ankles as well as glued-in waist elastic.
- Elasticated thumb loops prevent sleeves from riding up.
Specifications
Hooded Coveralls | |
Single-use | |
Elastic, Attached Socks, Boot Overflaps | |
Hooded | |
Zipper with Self-adhesive Flap and Chin Flap | |
Category III, Type 4-B, 5-B and 6-B | |
(EU) 2016/425, EN 14126, EN 1149-5 | |
80/Pk. |
HDPE | |
Elastic | |
Thumb Loop | |
Elastic | |
Stitched, Taped | |
Self-adhesive chin flap | |
For maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or in research and biosecurity laboratories, medical applications and when exposed to biological hazards |
The user should make the risk assessment to determine the protection required.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.