missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek 500 Jacket
268.64 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Specially designed for use with Tyvek™ apparel
- Flash-spun HDPE construction
- Stitched internal seams
- Front zip closure
- Elasticated face and wrists
- White
- PPE Category I
- Antistatic treatment (EN 1149-5) - on both sides
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | X-Large |
Material | HDPE |
Color | White |
Closure Type | Zipper |
Certifications/Compliance | EN 1149-5 |
Seam Type | Stitched (Internal) |
Product Type | Hooded Chemical-resistant Jacket |
For Use With (Application) | Pharmaceutical Handling, Chemical Protection, Lead and Asbestos Abatement/remediation, General Maintenance/operations, Spray Painting and General Clean-up, Amongst Many Others |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'