missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Coverall with Shoe Covers
Description
Composed of flash spun high-density polyethylene. Dupont™ Tyvek™ IsoClean™ Coverall with Shoe Covers is permeable to both air and water vapor, yet repels water-based liquids and aerosols. Offers an excellent barrier against fine particles and fibers (down to 1μ in size) and is ultra-low-linting.
- Coverall with hood
- White
- Clean-processed and sterile
- Stitched internal seams
- Set sleeve design with elastication at wrists, ankles and face
- Attached thumb loops
- Zipper closure
- Attached overshoes with slip-retardant soles
- Not CE certified as chemical protective clothing
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Product Type | Cleanroom Coveralls |
Size | 3X-Large |
Material | Tyvek™ |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Front Zipper with Storm Flap |
Seam Type | Serged |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'