Learn More
DuPont™ Tyvek 500 lab cot wuth zipper
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Zipper and Pockets are composed of flash spun high density polyethylene, providing an idea balance of protection, durability and comfort.
Brand: DuPont™ D13398984
272.80 GBP valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Description
- Silicon non-added
- With collar, zipper closure and 2 pockets
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up

Specifications
Lab Coat | |
HDPE | |
Zipper | |
Chemicals | |
Stitched (internal) | |
108 to 116 cm | |
Pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up, amongst many others |
X-Large | |
White | |
Elastic | |
2 | |
Straight cut | |
Antistatic treatment (EN 1149-5) | |
Zipper closure. 3 pockets. Elasticated cuffs (tunneled), Stitched internal seams |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.