Learn More
DuPont™ Tyvek 500 Laboratory Coat no pockets
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Press Studs are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
Brand: DuPont™ D13398884
206.22 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
This item is not returnable.
View return policy
Description
- Silicon non-added
- With 5 press stud closures
- Elasticated cuffs (not tunnelled)
- Stitched internal seams
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
Large | |
White | |
Elastic | |
EN 1149-5 |
HDPE | |
Press Studs | |
Stitched (internal) |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Content Correction
Product Title
DuPont™ Tyvek 500 Laboratory Coat no pockets > Size: Large
Spot an opportunity for improvement?Share a Content Correction