Learn More
Poulten Selfe™ U-Tube Viscometer
Suitable for transparent liquids. With UKAS certificate of calibration
Brand: Poulten Selfe™ 1619/07
This item is not returnable.
View return policy
Description
- PSL calibrated viscometers are supplied with UKAS certificates providing an official assurance of the competence of the calibration work carried out. UKAS certificates of calibration, which satisfy the requirements of ISO 9000, are issued by the PSL Calibration Laboratory, which is a National Measurement Accreditation Service accredited laboratory.
- Type BS/U, IP71.Suitable for transparent liquids. Nominal overall length 300mm. Approx. sample size 13mL to 40mL.
Specifications
U-Tube Viscometer | |
UKAS, ISO 9000 | |
18 mL |
600 to 3000 cSt | |
G |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Poulten Selfe™ U-Tube Viscometer > Size: G; Viscosity Range: 600 to 3000mm2/s
Spot an opportunity for improvement?Share a Content Correction