missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet UB-S-90
GREENER_CHOICEBIOPHARMA

Safe and approved storage of flammable hazardous substances in working areas

POA

Specifications

Color Light Gray
Material Steel
Product Type Safety storage under bench cabinet
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Surface Coating Powder-coated plain
View More Specs

Alert:

For indoor use only


Products 11
Product Code Brand Door Style Model Weight (Metric) Price Quantity & Availability  
Product Code Brand Door Style Model Weight (Metric) Price Quantity & Availability  
15974526
GREENER_CHOICEBIOPHARMA
View accessory products View Documents
asecos™
3047300135295
Sliding UB90.060.059.050.S 111 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15924506
GREENER_CHOICEBIOPHARMA
View accessory products View Documents
asecos™
3045800135308
Sliding UB90.060.089.050.S 148 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15904436
View accessory products View Documents
asecos™
3042100135340
Sliding UB90.060.110.050.S 180 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15944456
View accessory products View Documents
asecos™
3042800135337
Sliding UB90.060.110.050.2S 183.50 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15958062
GREENER_CHOICEBIOPHARMA
View accessory products View Documents
asecos™
3047200135286
Sliding UB90.060.059.S 123.30 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15904496
View accessory products View Documents
asecos™
3045700135299
Sliding UB90.060.089.S 163 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15924426
View accessory products View Documents
asecos™
3042000135327
Sliding UB90.060.110.S 193 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15994436
View accessory products View Documents
asecos™
3042700135311
Sliding UB90.060.110.2S 200.10 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15974396
View accessory products View Documents
asecos™
3040100135345
Sliding UB90.060.140.2S 244 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15927476
View accessory products View Documents
asecos™
3265800135377
Sliding UB90.080.059.060.S 150.50 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

15907476
View accessory products View Documents
asecos™
3265700135373
Sliding UB90.080.110.060.2S 244.50 kg
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Safe and approved storage of flammable hazardous substances in working areas

Function / construction

  • Robust construction and longevity: body made of powder-coated sheet steel
  • User-friendly: drawer remains open in any position, interior of drawer completely visible – access to all containers
  • Safe: drawer fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
  • No unauthorised use: drawer lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Certificates

  • Fire resistance 90 minutes
  • type tested in accordance with EN 14470-1
  • EN 16121/16122 conformity
  • GS certified
  • CE compliant

Warranty

Five years

Specifications

Specifications

Light Gray
Safety storage under bench cabinet
Powder-coated plain
Steel
Safe and approved storage of flammable hazardous substances in working areas
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
asecos™ Type 90 Safety Storage Under Bench Cabinet UB-S-90

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.