missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Solvent-resistant Stirred Cells
For applications requiring compatibility with solvents. Solvent-Resistant Stirred Cells are autoclavable with membrane in-place.
Brand: Merck XFUF07601
This item is not returnable.
View return policy
Description
For applications requiring compatibility with solvents, this Millipore stirred cell is available in two sizes. Certain aldehydes, ketones (e. g., acetone) and aliphatic ethers/esters may reduce the life of the fluorocarbon O-rings.Borosilicate glass cylinder and PTFE components for broad compatibility
Autoclavable with membrane in-place
Top plate opening provides access to contents without dismantling
Few components, so it's easy to clean and assemble
For 47 mm disc filters
Applications: Concentration and Buffer Exchange Involving Organic Solvents

Specifications
Autoclavable | |
Borosilicate Glass | |
Protein desalting, protein purification, protein concentration | |
Solvent resistant | |
Stirred Cell | |
300mL | |
76mm | |
11cm |
For 76 mm membranes | |
40 cm2 | |
Solvent-resistant Stirred Cell, for 76 mm membranes | |
1 Ea. | |
1 mL | |
165 mm | |
76 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Merck Solvent-resistant Stirred Cells > Diameter: 76mm
Spot an opportunity for improvement?Share a Content Correction