Learn More
Thermo Scientific™ Ultra Pure Water Distributors
Use Thermo Scientific™ Ultra Pure Water Distributors with all SDI systems.
Brand:  Thermo Scientific™ 50155463
 View more versions of this product
Product Code. 15590226
  • £182.24 / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ Ultra Pure Water Distributors with all SDI systems.
  • Plastic
  • Connectors allow connection of shut-off taps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Distribution manifold: 1 inlet, 2 outlets
Ultra Pure Water Distribution Manifold
PVC
170 mm
SDI System
1 x R 3/4 in. union nut; 2 x R3/4 in. male thread
280 mm
50 mm