missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Ultra Thin Polyester Laboratory Labels

Durable Freezerbondz™ Polyester Markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.

£91.60 - £96.25

Specifications

Adhesive Type Acrylic
Color Black/White
Material Polyester
For Use With (Equipment) BMP™51, and BMP™53 Label Printers
View More Specs

Products 2
Product Code Brand Product Type Size Price Quantity & Availability  
Product Code Brand Product Type Size Price Quantity & Availability  
17084377
View Documents
Brady™
M-83-492
Label/Cartridge 12.7 mm dia. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15481111
View Documents
Brady™
143319
Tape 7.62 m x 12.7 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

B-492 labels can be applied to frozen surfaces, including glass and polypropylene, that have been stored in liquid nitrogen when wrapped around itself by at least 1/8 inches. Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable. Good print smudge resistance, solvent resistance and high/low temperature performance of -196° to 100°C. This all-in-one cartridge includes label supply and R6400 Series ribbon.

  • Availble in Label Cartridges, or Tape
  • Colour Black on White
  • Brady Material B-492 Ultra Thin Polyester
Specifications

Specifications

Acrylic
Black/White
Polyester
BMP™51, and BMP™53 Label Printers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.