missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraComp eBeads™ Compensation Beads

Designed for use in compensation with all fluorochromes excited by ultraviolet, violet, blue, green, yellow-green, and red lasers. UltraComp eBeads™ react with antibodies of mouse, rat, and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test.

Brand:  Invitrogen™ 01-2222-42

200.20 GBP valid until 2025-02-13
Use promo code "24070" to get your promotional price.


 View more versions of this product

Product Code. 15385356

  • £286.00 / 100 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

Contains Sodium azide

This item is not returnable. View return policy

Description

Description

  • Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
  • This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
  • Compatible with all fluorochromes excited by an ultraviolet (355nm) or violet (405nm) laser
  • Should be used with standard staining buffers which contain PBS or HBSS, protein such as BSA or FBS, and sodium azide. No other additives should be used.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometer
100 tests
Compensation Beads
100 Tests
eBeads™
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2978462 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number01-2222-42
Lot Number2790074 Certificate TypeCertificate of Analysis Date06/11/2023 Catalog Number01-2222-42
Lot Number2743108 Certificate TypeCertificate of Analysis Date17/08/2023 Catalog Number01-2222-42
Lot Number2566360 Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number01-2222-42
Lot Number2563856 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog Number01-2222-42
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ UltraComp eBeads™ Compensation Beads > 100 tests

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.