missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraComp eBeads™ Compensation Beads

Product Code. 16881320
Click to view available options
Quantity:
100 tests
25 tests
Unit Size:
100 tests
25 tests
Each
This item is not returnable. View return policy
Contains Sodium azide

Product Code. 16881320

Brand: Invitrogen™ 01222242

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Contains Sodium azide

Designed for use in compensation with all fluorochromes excited by ultraviolet, violet, blue, green, yellow-green, and red lasers. UltraComp eBeads™ react with antibodies of mouse, rat, and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test.

  • Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
  • This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
  • Compatible with all fluorochromes excited by an ultraviolet (355nm) or violet (405nm) laser
  • Should be used with standard staining buffers which contain PBS or HBSS, protein such as BSA or FBS, and sodium azide. No other additives should be used.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Flow Cytometer
No. of Tests 100 Tests
Quantity 100 tests
Product Line eBeads
Product Type Compensation Beads

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.