missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UNC 0642
Potent and selective G9a and GLP inhibitor
£251.00 - £1035.00
Specifications
Purity | >99% |
---|---|
Content And Storage | -20°C |
Product Type | UNC 0642 |
Molecular Weight (g/mol) | 546.7 |
Inhibitors | UNC 0642 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18712449
|
Tocris Bioscience™
5132/10 |
10 mg |
£251.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18761819
|
Tocris Bioscience™
5132/50 |
50 mg |
£1035.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective G9a and GLP histone lysine methyltransferase inhibitor (IC50 < 2.5 nM). Exhibits >2,000-fold selectivity for G9a and GLP over PRC2-EZH2 and >20,000-fold selectivity over other methyltransferases. Reduces H3K9 dimethylation levels in MDA-MB-231 cells (IC50 = 110 nM). Displays modest brain penetration in vivo.Specifications
>99% | |
UNC 0642 | |
UNC 0642 |
-20°C | |
546.7 |
For Research Use Only