Learn More
Tocris Bioscience™ UNC 2327
Allosteric inhibitor of PRMT3
Brand:  Tocris Bioscience™ 5057/10
157.23 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18725758
  • £197.00 / 10mg
Explore more special offers
Description
Description
UNC 2327 is an allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 230 nM). Noncompetitive with both the peptide substrate and cofactor.
Specifications
Specifications
>99%
319.38
10 mg
UNC 2327
UNC 2327