missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UNC 2327
Allosteric inhibitor of PRMT3
£196.54 - £746.20
Specifications
Purity | >99% |
---|---|
Description | UNC 2327 |
Molecular Weight (g/mol) | 319.38 |
Product Type | UNC 2327 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725758
|
Tocris Bioscience™
5057/10 |
10 mg |
£196.54
10mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18747678
|
Tocris Bioscience™
5057/50 |
50 mg |
£746.20
50mg |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 230 nM). Noncompetitive with both the peptide substrate and cofactor.Specifications
>99% | |
UNC 2327 | |
319.38 | |
UNC 2327 |
For Research Use Only