Learn More
Sartorius™ Replacement Screen Kits for Use with Glass Holders for Vacuum Filtration Membranes
Replacement Screen Kits consist of a PTFE-coated, stainless steel drain screen and support screen together with a PTFE ring and silicone O-ring. They are intended for use with Series 16316 and 16315 Glass Holders for Vacuum Filtration Membranes.
Brand:  Sartorius™ 17147
 View more versions of this product
Product Code. 10586081
  • £232.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Product Type: Replacement Screen Kits
  • Material: PTFE (Main Ring), PTFE/Stainless Steel (Drain Screen, Support Screen), Silicone (O-Ring)
  • Includes: Support Screen, Drain Screen, Main Ring, O-Ring
  • For Use With (Equipment): Sartorius™ Glass Holders for Vacuum Filtration Membranes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Filter Holder
Non-sterile
Series 16316 Glass Holders for Vacuum Filtration Membranes
17147
50 mm/47 mm
PTFE (Ring), PTFE/Stainless Steel (Drain Screen, Support Screen), Silicone (O-ring)
Vacuum Filtration
Main Ring, Drain Screen, Support Screen, O-Ring
Screen Kit