Learn More
Köttermann Underbench Acid and Base Cabinet
Constructed with durable polypropylene, this Underbench Acid and Base Cabinet is built to endure the aggressive conditions produced by corrosive acid fumes. Liquid-tight pull-out trays offer easy access while minimizing health and safety risks.
Brand:  Köttermann 377-0005
Additional Details : Weight : 43.00000kg
 View more versions of this product
Product Code. 17332159
  • £1192.50 / Each
Estimated Shipment: 05-11-2024
to see stock.
Description
Description
  • Safe storage of corrosive non-flammable hazardous substances, with acids and bases separated
  • Cabinet carcasses and pull-out trays made of resistant polypropylene
  • Liquid-tight drawers on low-friction PTFE strips with pull-out stop
  • Removal of aggressive vapors via optional ventilation with an external fan
  • Corrosion-resistant locks and hinges
  • Ideal for installation under high-performance fume cupboards
Specifications
Specifications
No Ventilation Fan
White
Acid and Base Storage
Polypropylene
515 x 600 x 720 mm
1 Left-hinged Door, 2 Drawers
Hinged, Drawer