missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Köttermann Underbench Acid and Base Cabinet
Constructed with durable polypropylene, this Underbench Acid and Base Cabinet is built to endure the aggressive conditions produced by corrosive acid fumes. Liquid-tight pull-out trays offer easy access while minimizing health and safety risks.
£951.88 - £3323.10
Specifications
Color | White |
---|---|
Material | Polypropylene |
Door Style | Hinged, Drawer |
For Use With (Application) | Acid and Base Storage |
Product Code | Brand | Dimensions (D x W x H) | Configuration | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (D x W x H) | Configuration | Includes | Price | Quantity & Availability | |||
17302179
|
Köttermann
377-0022 |
520 x 450 x 720 mm | 1 Right-hinged Door, 2 Pull-outs | No Ventilation Fan |
£1351.50
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17322179
|
Köttermann
377-0024 |
520 x 450 x 720 mm | 1 Right-hinged Door, 1 Pull-out | No Ventilation Fan |
£951.88
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17332169
|
Köttermann
377-0015 |
515 x 600 x 720 mm | 1 Left-hinged Door, 1 Drawer for Containers | No Ventilation Fan |
£1287.90
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17332159
|
Köttermann
377-0005 |
515 x 600 x 720 mm | 1 Left-hinged Door, 2 Drawers | No Ventilation Fan |
£1192.50
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17342169
|
Köttermann
377-0016 |
515 x 600 x 720 mm | 1 Right-hinged Door, 1 Drawer for Containers | No Ventilation Fan |
£1287.90
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17342159
|
Köttermann
377-0006 |
515 x 600 x 720 mm | 1 Right-hinged Door, 2 Drawers | No Ventilation Fan |
£1192.50
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17392159
|
Köttermann
377-0011 |
515 x 900 x 720 mm | 2 Doors, 3 Drawers | Ventilation Fan |
£2946.80
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17352169
|
Köttermann
377-0017 |
515 x 900 x 720 mm | 2 Doors, 3 Drawers (1 for Containers) | No Ventilation Fan |
£2045.80
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17352159
|
Köttermann
377-0007 |
515 x 900 x 720 mm | 2 Doors, 4 Drawers | No Ventilation Fan |
£2109.40
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17302169
|
Köttermann
377-0012 |
515 x 1100 x 720 mm | 2 Doors, 3 Drawers | Ventilation Fan |
£3015.70
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17362169
|
Köttermann
377-0018 |
515 x 1100 x 720 mm | 2 Doors, 3 Drawers (1 for Containers) | No Ventilation Fan |
£2109.40
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17362159
|
Köttermann
377-0008 |
515 x 1100 x 720 mm | 2 Doors, 4 Drawers | No Ventilation Fan |
£2178.30
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17312169
|
Köttermann
377-0013 |
515 x 1200 x 720 mm | 2 Doors, 3 Drawers | Ventilation Fan |
£3079.30
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17372169
|
Köttermann
377-0019 |
515 x 1200 x 720 mm | 2 Doors, 3 Drawers (1 for Containers) | No Ventilation Fan |
£2178.30
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17372159
|
Köttermann
377-0009 |
515 x 1200 x 720 mm | 2 Doors, 4 Drawers | No Ventilation Fan |
£2252.50
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17322169
|
Köttermann
377-0014 |
515 x 1400 x 720 mm | 2 Doors, 3 Drawers | Ventilation Fan |
£3323.10
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17382169
|
Köttermann
377-0020 |
515 x 1400 x 720 mm | 2 Doors, 3 Drawers (1 for Containers) | No Ventilation Fan |
£2427.40
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17382159
|
Köttermann
377-0010 |
515 x 1400 x 720 mm | 2 Doors, 4 Drawers | No Ventilation Fan |
£2496.30
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17312179
|
Köttermann
377-0023 |
520 x 450 x 720 mm | 1 Left-hinged Door, 1 Pull-out | No Ventilation Fan |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
17392169
|
Köttermann
377-0021 |
520 x 450 x 720 mm | 1 Left-hinged Door, 2 Pull-outs | No Ventilation Fan |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Safe storage of corrosive non-flammable hazardous substances, with acids and bases separated
- Cabinet carcasses and pull-out trays made of resistant polypropylene
- Liquid-tight drawers on low-friction PTFE strips with pull-out stop
- Removal of aggressive vapors via optional ventilation with an external fan
- Corrosion-resistant locks and hinges
- Ideal for installation under high-performance fume cupboards
Specifications
White | |
Polypropylene | |
Hinged, Drawer | |
Acid and Base Storage |