Learn More
Duran™ Ungraduated Measuring Cylinders
These measuring cylinders are for holding and measuring varying amounts of liquids in the laboratory.
Brand: Duran™ 213912477
This item is not returnable.
View return policy
Description
- Without graduation
- Volume content tolerances for measuring and mixing cylinders conform to DIN and ISO accuracy limits
- Robust, virtually inert and exceptionally resistant to high temperatures
- Outstanding corrosion resistance
- Manufactured from borosilicate glass 3.3
- Large hexagonal base has three knobs to increase stability and prevents cylinder from rolling
Specifications
Laboratories | |
Ungraduated Measuring Cylinder | |
Borosilicate Glass | |
DIN EN ISO 4788 |
Hexagonal | |
100 mL | |
10/pk |