Learn More
Abeba™ UNI6 36781 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Brand:  Abeba™ 36781/47
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15610668
  • £90.81 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRC
  • Microfiber black
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRC
Microfiber
Slip-resistant
36781
Black
47
Polyurethane