Learn More
Abeba™ UNI6 36781 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
£90.81
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 O2 FO SRC |
---|---|
Color | Black |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15690648
|
Abeba™
36781/35 |
35 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15600658
|
Abeba™
36781/36 |
36 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15610658
|
Abeba™
36781/37 |
37 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15620658
|
Abeba™
36781/38 |
38 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15630658
|
Abeba™
36781/39 |
39 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15640658
|
Abeba™
36781/40 |
40 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15650658
|
Abeba™
36781/41 |
41 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15660658
|
Abeba™
36781/42 |
42 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15670658
|
Abeba™
36781/43 |
43 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15680658
|
Abeba™
36781/44 |
44 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15690658
|
Abeba™
36781/45 |
45 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15600668
|
Abeba™
36781/46 |
46 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15610668
|
Abeba™
36781/47 |
47 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15620668
|
Abeba™
36781/48 |
48 |
£90.81
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 O2 FO SRC
- Microfiber black
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable Soft Comfort insole M (article no. 350112)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
Specifications
CE EN ISO 20347:2012 O2 FO SRC | |
Microfiber | |
Polyurethane |
Black | |
Slip-resistant | |
36781 |