Learn More
Alexandra™ Unisex Scrub Trousers, Short, Caribbean Blue Product Code.: 15880664
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
Alexandra™ Unisex Scrub Trousers, Short, Caribbean Blue
Product Code. 15880664
3X-Large, Short
Click to view available options
Size:
2X-Large, Short
3X-Large, Short
4X-Large, Short
Medium, Short
Short
X-Large, Short
X-Small, Short
Product Code. 15880664
Brand: Alexandra™ D398CA3XLS
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
Add to basket
Unisex scrub trousers suitable for a professional healthcare uniform. Lightweight hardwearing and practical with one back pocket. Available in a range of colors.
  • Elasticated drawstring waist
  • Machine washable and industrially launderable
  • Inside leg: Short (S) 29 in. Regular (R) 31 in. Tall (T) 33 in. Unhemmed (U) 38in.
  • Fabric Weight: 145gsm
  • 67% polyester/33% cotton
Specifications
Size 3X-Large, Short
Color Caribbean Blue
Style D398
Product Content Correction
Product Title
Alexandra™ Unisex Scrub Trousers, Short, Caribbean Blue >