Learn More
Fisherbrand™ X3 Universal Indicator pH Paper Roll Refill, pH 1-14
Quickly and easily measure solution pH value at the point of interest with this paper impregnated with universal indicator solution. Fisherbrand™ Universal Indicator pH Paper Roll exhibits smooth color changes over a pH value range from 1 to 14.
Brand:  Fisherbrand™ 0911Z00035
Product Code. 10151622
  • £20.92 / Pack of 3
Edge
Explore more special offers
Description
Description
  • Ready to use.
  • No calibration, set-up, or maintenance.
  • Perfect for a variety of environments.
Order Info
This is a refill for Fisherbrand Universal pH Indicator Paper, Range 1-14 (Code 11763554).
Specifications
Specifications
1 to 14
1 Roll
Universal pH Indicator Paper - Refill, grade 911
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ X3 Universal Indicator pH Paper Roll Refill, pH 1-14 > Quantity: 1 roll