Learn More
Thermo Scientific Chemicals uPA Inhibitor II, UK122
Marke: Thermo Scientific Chemicals J64874.#0
Beschreibung
A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activatoruPA Inhibitor II, UK122 acts as a potent, selective and active-site targeting urokinase-type plasminogen activator (uPA) inhibitor. However, its activity against trypsin, thrombin, tPA, and plasmin is greatly reduced. It is also effectively inhibits the migration (80% inhibition) and invasion (68% inhibition) of uPA-expressing CFPAC-1 pancreatic cells, without affecting its proliferation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator
Spezifikation
5 mg | |
2°C to 8°C | |
C17H13N3O2·CF3CO2H | |
Light Sensitive | |
Solid |
Yellow | |
A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator | |
405.30g/mol | |
Signal Transduction Reagent |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
RUO – Research Use Only