Learn More
Thermo Scientific Chemicals uPA Inhibitor II, UK122
£143.72 - £559.79
Specifications
Description | Yellow |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator |
Molecular Formula | C17H13N3O2·CF3CO2H |
Formula Weight | 405.30g/mol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15496619
|
Thermo Scientific Chemicals
J64874.MB |
25 mg |
£559.79
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15486619
|
Thermo Scientific Chemicals
J64874.#0 |
5 mg |
£143.72
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activatoruPA Inhibitor II, UK122 acts as a potent, selective and active-site targeting urokinase-type plasminogen activator (uPA) inhibitor. However, its activity against trypsin, thrombin, tPA, and plasmin is greatly reduced. It is also effectively inhibits the migration (80% inhibition) and invasion (68% inhibition) of uPA-expressing CFPAC-1 pancreatic cells, without affecting its proliferation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator
Specifications
Yellow | |
A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator | |
405.30g/mol | |
Signal Transduction Reagent |
2°C to 8°C | |
C17H13N3O2·CF3CO2H | |
Light Sensitive | |
Solid |
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.