Learn More
Kinesis™ Adapters for HPLC
Convenience of quick connect and disconnect of a luer connection
Brand: Kinesis™ P-604
This item is not returnable.
View return policy
Description
Convenience of quick connect and disconnect of a luer connection
- Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Threaded Luer Adapter | |
Female internal threads Luer to 1/4-28 Flat-bottom | |
Acetal | |
Female Luer x Male 1/4-28 Flat Bottom |